Anti-PKC zeta Rabbit Polyclonal Antibody [clone: P556]
Lieferant: Bioworld Technology
BS1579
BWRLBS1579EA 262 EUR
BWRLBS1579
Anti-PKC zeta Rabbit Polyclonal Antibody [clone: P556]
Antikörper
Synthetic peptide, corresponding to amino acids 520-570 of Human PKC zeta.
PKC ? (P556) pAb detects endogenous levels of PKC ? protein.
Type: Primary
Antigen: PKC zeta
Clonality: Polyclonal
Clone: P556
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat