Anti-PKC-pan Rabbit Polyclonal Antibody [clone: T495]

Lieferant: Bioworld Technology
BS1580
BWRLBS1580EA 262 EUR
BWRLBS1580
Anti-PKC-pan Rabbit Polyclonal Antibody [clone: T495]
Antikörper
Synthetic peptide, corresponding to amino acids 470-520 of Human PKC-pan.

PKC-pan (T495) pAb detects endogenous levels of PKC ?, ? I, ? II, ?, ?, ?, ?, ? and ? isoforms.

Type: Primary
Antigen: PKC-pan
Clonality: Polyclonal
Clone: T495
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR