Anti-PKD1 Rabbit Polyclonal Antibody [clone: G459]
Lieferant: Bioworld Technology
BS1581
BWRLBS1581EA 262 EUR
BWRLBS1581
Anti-PKD1 Rabbit Polyclonal Antibody [clone: G459]
Antikörper
Synthetic peptide, corresponding to amino acids 424-482 of Human PKD1.
PKD1 (G459) pAb detects endogenous levels of PKD1 protein.
Type: Primary
Antigen: PKD1
Clonality: Polyclonal
Clone: G459
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat