Anti-SF1 Rabbit Polyclonal Antibody [clone: P76]
Lieferant: Bioworld Technology
BS1593
BWRLBS1593EA 262 EUR
BWRLBS1593
Anti-SF1 Rabbit Polyclonal Antibody [clone: P76]
Antikörper
Synthetic peptide, corresponding to amino acids 45-98 of Human SF1.
SF1 (P76) pAb detects endogenous levels of SF1 protein.
Type: Primary
Antigen: SF1
Clonality: Polyclonal
Clone: P76
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse