Anti-XIAP Rabbit Polyclonal Antibody [clone: G81]

Lieferant: Bioworld Technology
BS1609
BWRLBS1609EA 262 EUR
BWRLBS1609
Anti-XIAP Rabbit Polyclonal Antibody [clone: G81]
Antikörper
Synthetic peptide, corresponding to amino acids 50-100 of Human XIAP.

XIAP (G81) pAb detects endogenous levels of XIAP protein.

Type: Primary
Antigen: XIAP
Clonality: Polyclonal
Clone: G81
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR