Anti-AURKB Rabbit Polyclonal Antibody [clone: L228]

Lieferant: Bioworld Technology
BS1613
BWRLBS1613EA 262 EUR
BWRLBS1613
Anti-AURKB Rabbit Polyclonal Antibody [clone: L228]
Antikörper
Synthetic peptide, corresponding to amino acids 194-245 of Human AURKB.

AURKB (L228) pAb detects endogenous levels of AURKB protein.

Type: Primary
Antigen: AURKB
Clonality: Polyclonal
Clone: L228
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR