Anti-Tubulin alpha Rabbit Polyclonal Antibody [clone: G436]

Lieferant: Bioworld Technology
BS1699
BWRLBS1699EA 262 EUR
BWRLBS1699
Anti-Tubulin alpha Rabbit Polyclonal Antibody [clone: G436]
Antikörper
Synthetic peptide, corresponding to amino acids 201-256 of Human Tubulin alpha.

Tubulin ? (G436) pAb detects endogenous levels of Tubulin ? protein.

Type: Primary
Antigen: Tubulin alpha
Clonality: Polyclonal
Clone: G436
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR