Anti-IL-2Ralpha Rabbit Polyclonal Antibody [clone: R264]
Lieferant: Bioworld Technology
BS1758
BWRLBS1758EA 262 EUR
BWRLBS1758
Anti-IL-2Ralpha Rabbit Polyclonal Antibody [clone: R264]
Antikörper
Synthetic peptide, corresponding to amino acids 231-280 of Human IL-2Ralpha.
IL-2R? (R264) pAb detects endogenous levels of IL-2R? protein.
Type: Primary
Antigen: IL-2Ralpha
Clonality: Polyclonal
Clone: R264
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human