Anti-PR Rabbit Polyclonal Antibody [clone: A394]

Lieferant: Bioworld Technology
BS1766
BWRLBS1766EA 262 EUR
BWRLBS1766
Anti-PR Rabbit Polyclonal Antibody [clone: A394]
Antikörper
Synthetic peptide, corresponding to amino acids 361-410 of Human PR.

PR (A394) pAb detects endogenous levels of PR protein.

Type: Primary
Antigen: PR
Clonality: Polyclonal
Clone: A394
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR