Anti-14-3-3 Theta Rabbit Polyclonal Antibody [clone: W228]
Lieferant: Bioworld Technology
BS1790
BWRLBS1790EA 262 EUR
BWRLBS1790
Anti-14-3-3 Theta Rabbit Polyclonal Antibody [clone: W228]
Antikörper
Synthetic peptide, corresponding to amino acids 200-240 of Human 14-3-3 Theta.
14-3-3 ? (W228) pAb detects endogenous levels of 14-3-3 protein theta/tau.
Type: Primary
Antigen: 14-3-3 Theta
Clonality: Polyclonal
Clone: W228
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat