Anti-AhR Rabbit Polyclonal Antibody [clone: K32]

Lieferant: Bioworld Technology
BS1791
BWRLBS1791EA 262 EUR
BWRLBS1791
Anti-AhR Rabbit Polyclonal Antibody [clone: K32]
Antikörper
Synthetic peptide, corresponding to amino acids N-terminus of Human AhR.

AhR (K32) pAb detects endogenous levels of AhR protein.

Type: Primary
Antigen: AhR
Clonality: Polyclonal
Clone: K32
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR