Anti-EGFR Rabbit Polyclonal Antibody [clone: F1065]
Lieferant: Bioworld Technology
BS1795
BWRLBS1795EA 262 EUR
BWRLBS1795
Anti-EGFR Rabbit Polyclonal Antibody [clone: F1065]
Antikörper
Synthetic peptide, corresponding to amino acids 1041-1090 of Human EGFR.
EGFR (F1065) pAb detects endogenous levels of EGFR protein.
Type: Primary
Antigen: EGFR
Clonality: Polyclonal
Clone: F1065
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat