Anti-ErbB-4 Rabbit Polyclonal Antibody [clone: E1280]

Lieferant: Bioworld Technology
BS1797
BWRLBS1797EA 262 EUR
BWRLBS1797
Anti-ErbB-4 Rabbit Polyclonal Antibody [clone: E1280]
Antikörper
Synthetic peptide, corresponding to amino acids 1246-1310 of Human ErbB-4.

ErbB-4 (E1280) pAb detects endogenous levels of ErbB-4 protein.

Type: Primary
Antigen: ErbB-4
Clonality: Polyclonal
Clone: E1280
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR