Anti-Integrin beta4 Rabbit Polyclonal Antibody [clone: L1506]

Lieferant: Bioworld Technology
BS1802
BWRLBS1802EA 262 EUR
BWRLBS1802
Anti-Integrin beta4 Rabbit Polyclonal Antibody [clone: L1506]
Antikörper
Synthetic peptide, corresponding to amino acids 1470-1520 of Human Integrin beta4.

Integrin ?4 (L1506) pAb detects endogenous levels of Integrin ?4 protein.

Type: Primary
Antigen: Integrin beta4
Clonality: Polyclonal
Clone: L1506
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR