Anti-PDCD4 Rabbit Polyclonal Antibody [clone: K453]

Lieferant: Bioworld Technology
BS1805
BWRLBS1805EA 262 EUR
BWRLBS1805
Anti-PDCD4 Rabbit Polyclonal Antibody [clone: K453]
Antikörper
Synthetic peptide, corresponding to amino acids 422-474 of Human PDCD4.

PDCD4 (K453) pAb detects endogenous levels of PDCD4 protein.

Type: Primary
Antigen: PDCD4
Clonality: Polyclonal
Clone: K453
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR