Anti-AKT Rabbit Polyclonal Antibody [clone: A444]

Lieferant: Bioworld Technology
BS1810
BWRLBS1810EA 262 EUR
BWRLBS1810
Anti-AKT Rabbit Polyclonal Antibody [clone: A444]
Antikörper
Synthetic peptide, corresponding to amino acids 420-470 of Human AKT1.

AKT (A444) pAb detects endogenous levels of total AKT protein.

Type: Primary
Antigen: AKT
Clonality: Polyclonal
Clone: A444
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR