Anti-AKT1 Rabbit Polyclonal Antibody [clone: M118]
Lieferant: Bioworld Technology
BS1811
BWRLBS1811EA 262 EUR
BWRLBS1811
Anti-AKT1 Rabbit Polyclonal Antibody [clone: M118]
Antikörper
Synthetic peptide, corresponding to amino acids 90-130 of Human AKT1.
AKT1 (M118) pAb detects endogenous levels of AKT1 protein, this antibody does not cross-react with AKT2 or AKT3.
Type: Primary
Antigen: AKT1
Clonality: Polyclonal
Clone: M118
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat