Anti-IKKalpha/beta Rabbit Polyclonal Antibody [clone: D173]
Lieferant: Bioworld Technology
BS1815
BWRLBS1815EA 262 EUR
BWRLBS1815
Anti-IKKalpha/beta Rabbit Polyclonal Antibody [clone: D173]
Antikörper
Synthetic peptide, corresponding to amino acids 151-200 of Human IKKalpha
IKK?/? (D173) pAb detects endogenous levels of IKK?/? protein.
Type: Primary
Antigen: IKKalpha/beta
Clonality: Polyclonal
Clone: D173
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat