Anti-Ku-70 Rabbit Polyclonal Antibody [clone: S1]

Lieferant: Bioworld Technology
BS1818
BWRLBS1818EA 262 EUR
BWRLBS1818
Anti-Ku-70 Rabbit Polyclonal Antibody [clone: S1]
Antikörper
Synthetic peptide, corresponding to the N-terminus of Human Ku-70.

Ku70 (S1) pAb detects endogenous levels of Ku70 protein.

Type: Primary
Antigen: Ku-70
Clonality: Polyclonal
Clone: S1
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR