Anti-14-3-3 zeta Rabbit Polyclonal Antibody [clone: W228]

Lieferant: Bioworld Technology
BS1826
BWRLBS1826EA 262 EUR
BWRLBS1826
Anti-14-3-3 zeta Rabbit Polyclonal Antibody [clone: W228]
Antikörper
Synthetic peptide, corresponding to amino acids 200-250 of Human 14-3-3 zeta.

14-3-3 ? (W228) pAb detects endogenous levels of 14-3-3 protein zeta/delta.

Type: Primary
Antigen: 14-3-3 zeta
Clonality: Polyclonal
Clone: W228
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR