Anti-Cdc25A Rabbit Polyclonal Antibody [clone: K120]

Lieferant: Bioworld Technology
BS1831
BWRLBS1831EA 262 EUR
BWRLBS1831
Anti-Cdc25A Rabbit Polyclonal Antibody [clone: K120]
Antikörper
Synthetic peptide, corresponding to amino acids 280-340 of Human Cdc25A.

Cdc25A (K120) pAb detects endogenous levels of Cdc25A protein.

Type: Primary
Antigen: Cdc25A
Clonality: Polyclonal
Clone: K120
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR