Anti-Di-Ras1 Rabbit Polyclonal Antibody [clone: R181]

Lieferant: Bioworld Technology
BS1897
BWRLBS1897EA 262 EUR
BWRLBS1897
Anti-Di-Ras1 Rabbit Polyclonal Antibody [clone: R181]
Antikörper
Synthetic peptide, corresponding to amino acids 151-200 of Human Di-Ras1.

Di-Ras1 (R181) pAb detects endogenous levels of Di-Ras1 protein.

Type: Primary
Antigen: Di-Ras1
Clonality: Polyclonal
Clone: R181
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR