Anti-VEGI Rabbit Polyclonal Antibody [clone: F141]

Lieferant: Bioworld Technology
BS1899
BWRLBS1899EA 262 EUR
BWRLBS1899
Anti-VEGI Rabbit Polyclonal Antibody [clone: F141]
Antikörper
Synthetic peptide, corresponding to amino acids 111-160 of Human VEGI.

VEGI (F141) pAb detects endogenous levels of VEGI protein.

Type: Primary
Antigen: VEGI
Clonality: Polyclonal
Clone: F141
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR