Anti-UMP-CMP Kinase Rabbit Polyclonal Antibody [clone: E36]

Lieferant: Bioworld Technology
BS1975
BWRLBS1975EA 262 EUR
BWRLBS1975
Anti-UMP-CMP Kinase Rabbit Polyclonal Antibody [clone: E36]
Antikörper
Synthetic peptide, corresponding to the N-terminal of Human UMP-CMP Kinase.

UMP-CMP Kinase (E36) pAb detects endogenous levels of UMP-CMP Kinase protein.

Type: Primary
Antigen: UMP-CMP Kinase
Clonality: Polyclonal
Clone: E36
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

TECHNISCHE DATEN

Antigen-Symbol UMP-CMP Kinase
Antikörper-Typ Primär
Klonalität Polyklonal
Klon E36
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Western blot Yes
ImmunoChemie Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR