Anti-Hox-A1 Rabbit Polyclonal Antibody [clone: Q203]

Lieferant: Bioworld Technology
BS2003
BWRLBS2003EA 262 EUR
BWRLBS2003
Anti-Hox-A1 Rabbit Polyclonal Antibody [clone: Q203]
Antikörper
Synthetic peptide, corresponding to amino acids 180-220 of Human Hox-A1.

Hox-A1 (Q203) pAb detects endogenous levels of Hox-A1 protein.

Type: Primary
Antigen: Hox-A1
Clonality: Polyclonal
Clone: Q203
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR