Anti-GK2 Rabbit Polyclonal Antibody [clone: Y306]

Lieferant: Bioworld Technology
BS2060
BWRLBS2060EA 262 EUR
BWRLBS2060
Anti-GK2 Rabbit Polyclonal Antibody [clone: Y306]
Antikörper
Synthetic peptide, corresponding to amino acids 276-325 of Human GK2.

GK2 (Y306) pAb detects endogenous levels of GK2 protein.

Type: Primary
Antigen: GK2
Clonality: Polyclonal
Clone: Y306
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR