Anti-C/EBP-epsilon Rabbit Polyclonal Antibody [clone: K220]
Lieferant: Bioworld Technology
BS2074
BWRLBS2074EA 262 EUR
BWRLBS2074
Anti-C/EBP-epsilon Rabbit Polyclonal Antibody [clone: K220]
Antikörper
Synthetic peptide, corresponding to amino acids 188-245 of Human C/EBP-epsilon.
C/EBP-? (K220) pAb detects endogenous levels of C/EBP-? protein.
Type: Primary
Antigen: C/EBP-epsilon
Clonality: Polyclonal
Clone: K220
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat