Anti-Septin 1 Rabbit Polyclonal Antibody [clone: D205]

Lieferant: Bioworld Technology
BS2111
BWRLBS2111EA 262 EUR
BWRLBS2111
Anti-Septin 1 Rabbit Polyclonal Antibody [clone: D205]
Antikörper
Synthetic peptide, corresponding to amino acids 169-224 of Human Septin 1.

Septin 1 (D205) pAb detects endogenous levels of Septin 1 protein.

Type: Primary
Antigen: Septin 1
Clonality: Polyclonal
Clone: D205
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR