Anti-SMIF Rabbit Polyclonal Antibody [clone: S133]
Lieferant: Bioworld Technology
BS2145
BWRLBS2145EA 262 EUR
BWRLBS2145
Anti-SMIF Rabbit Polyclonal Antibody [clone: S133]
Antikörper
Synthetic peptide, corresponding to amino acids 102-150 of Human SMIF.
SMIF (S133) pAb detects endogenous levels of SMIF protein.
Type: Primary
Antigen: SMIF
Clonality: Polyclonal
Clone: S133
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse