Anti-PERK Rabbit Polyclonal Antibody [clone: R87]
Lieferant: Bioworld Technology
BS2156
BWRLBS2156EA 262 EUR
BWRLBS2156
Anti-PERK Rabbit Polyclonal Antibody [clone: R87]
Antikörper
Synthetic peptide, corresponding to amino acids 56-102 of Human PERK.
PERK (R87) pAb detects endogenous levels of PERK protein.
Type: Primary
Antigen: PERK
Clonality: Polyclonal
Clone: R87
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human