Anti-FoxD3 Rabbit Polyclonal Antibody [clone: R233]
Lieferant: Bioworld Technology
BS2163
BWRLBS2163EA 262 EUR
BWRLBS2163
Anti-FoxD3 Rabbit Polyclonal Antibody [clone: R233]
Antikörper
Synthetic peptide, corresponding to amino acids 202-251 of Human FoxD3.
FoxD3 (R233) pAb detects endogenous levels of FoxD3 protein.
Type: Primary
Antigen: FoxD3
Clonality: Polyclonal
Clone: R233
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat