Anti-RAGE Rabbit Polyclonal Antibody [clone: Q286]

Lieferant: Bioworld Technology
BS2165
BWRLBS2165EA 262 EUR
BWRLBS2165
Anti-RAGE Rabbit Polyclonal Antibody [clone: Q286]
Antikörper
Synthetic peptide, corresponding to amino acids 254-308 of Human RAGE.

RAGE (Q286) pAb detects endogenous levels of RAGE protein.

Type: Primary
Antigen: RAGE
Clonality: Polyclonal
Clone: Q286
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR