Anti-Cdc37 Rabbit Polyclonal Antibody [clone: F259]
Lieferant: Bioworld Technology
BS2201
BWRLBS2201EA 262 EUR
BWRLBS2201
Anti-Cdc37 Rabbit Polyclonal Antibody [clone: F259]
Antikörper
Synthetic peptide, corresponding to amino acids 226-276 of Human Cdc37.
Cdc37 (F259) pAb detects endogenous levels of Cdc37 protein.
Type: Primary
Antigen: Cdc37
Clonality: Polyclonal
Clone: F259
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat