Anti-DNA Ligase IV Rabbit Polyclonal Antibody [clone: G619]

Lieferant: Bioworld Technology
BS2234
BWRLBS2234EA 262 EUR
BWRLBS2234
Anti-DNA Ligase IV Rabbit Polyclonal Antibody [clone: G619]
Antikörper
Synthetic peptide, corresponding to amino acids 600-650 of Human DNA Ligase IV.

DNA Ligase IV (G619) pAb detects endogenous levels of DNA Ligase IV protein.

Type: Primary
Antigen: DNA Ligase IV
Clonality: Polyclonal
Clone: G619
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR