Anti-RORalpha Rabbit Polyclonal Antibody [clone: G41]

Lieferant: Bioworld Technology
BS2272
BWRLBS2272EA 262 EUR
BWRLBS2272
Anti-RORalpha Rabbit Polyclonal Antibody [clone: G41]
Antikörper
Synthetic peptide, corresponding to amino acids 20-70 of Human RORalpha.

ROR? (G41) pAb detects endogenous levels of ROR? protein.

Type: Primary
Antigen: RORalpha
Clonality: Polyclonal
Clone: G41
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR