Anti-LTbetaR Rabbit Polyclonal Antibody [clone: R44]

Lieferant: Bioworld Technology
BS2273
BWRLBS2273EA 262 EUR
BWRLBS2273
Anti-LTbetaR Rabbit Polyclonal Antibody [clone: R44]
Antikörper
Synthetic peptide, corresponding to amino acids 20-60 of Human LTbetaR.

LT?R (R44) pAb detects endogenous levels of LT?R protein.

Type: Primary
Antigen: LTbetaR
Clonality: Polyclonal
Clone: R44
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR