Anti-BUB3 Rabbit Polyclonal Antibody [clone: H274]

Lieferant: Bioworld Technology
BS2395
BWRLBS2395EA 262 EUR
BWRLBS2395
Anti-BUB3 Rabbit Polyclonal Antibody [clone: H274]
Antikörper
Synthetic peptide, corresponding to amino acids 251-300 of Human BUB3.

BUB3 (H274) pAb detects endogenous levels of BUB3 protein.

Type: Primary
Antigen: BUB3
Clonality: Polyclonal
Clone: H274
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

TECHNISCHE DATEN

Antigen-Symbol BUB3
Environmentally Preferable
Antikörper-Typ Primär
Klonalität Polyklonal
Klon H274
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Isotyp
Western blot Yes
ImmunoChemie Yes
ImmunFluoreszenz Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR