Anti-GAL9 Rabbit Polyclonal Antibody [clone: G80]
Lieferant: Bioworld Technology
BS2417
BWRLBS2417EA 262 EUR
BWRLBS2417
Anti-GAL9 Rabbit Polyclonal Antibody [clone: G80]
Antikörper
Synthetic peptide, corresponding to amino acids 51-100 of Human Galectin-9.
Galectin-9 (G80) pAb detects endogenous levels of Galectin-9 protein.
Type: Primary
Antigen: GAL9
Clonality: Polyclonal
Clone: G80
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat