Anti-TIF1alpha Rabbit Polyclonal Antibody [clone: Q1034]

Lieferant: Bioworld Technology
BS2449
BWRLBS2449EA 262 EUR
BWRLBS2449
Anti-TIF1alpha Rabbit Polyclonal Antibody [clone: Q1034]
Antikörper
Synthetic peptide, corresponding to amino acids 1011-1060 of Human TIF1alpha.

TIF1? (Q1034) pAb detects endogenous levels of TIF1? protein.

Type: Primary
Antigen: TIF1alpha
Clonality: Polyclonal
Clone: Q1034
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR