Anti-Exo1 Rabbit Polyclonal Antibody [clone: K86]

Lieferant: Bioworld Technology
BS2536
BWRLBS2536EA 262 EUR
BWRLBS2536
Anti-Exo1 Rabbit Polyclonal Antibody [clone: K86]
Antikörper
Synthetic peptide, corresponding to amino acids 55-05 of Human Exo1.

Exo1 (K86) pAb detects endogenous levels of Exo1 rotein.

Type: Primary
Antigen: Exo1
Clonality: Polyclonal
Clone: K86
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR