Anti-P-cadherin Rabbit Polyclonal Antibody [clone: G78]

Lieferant: Bioworld Technology
BS2577
BWRLBS2577EA 262 EUR
BWRLBS2577
Anti-P-cadherin Rabbit Polyclonal Antibody [clone: G78]
Antikörper
Synthetic peptide, corresponding to amino acids 45-90 of Human P-cadherin.

P-cadherin (G78) pAb detects endogenous levels of cadherin protein only when phosphorylated at Gly78.

Type: Primary
Antigen: P-cadherin
Clonality: Polyclonal
Clone: G78
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR