Anti-TACC3 Rabbit Polyclonal Antibody [clone: T813]

Lieferant: Bioworld Technology
BS2580
BWRLBS2580EA 262 EUR
BWRLBS2580
Anti-TACC3 Rabbit Polyclonal Antibody [clone: T813]
Antikörper
Synthetic peptide, corresponding to amino acids 790-830 of Human TACC3.

TACC3 (T813) pAb detects endogenous levels of TACC3 protein.

Type: Primary
Antigen: TACC3
Clonality: Polyclonal
Clone: T813
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR