Anti-EDG-5 Rabbit Polyclonal Antibody [clone: L303]

Lieferant: Bioworld Technology
BS2594
BWRLBS2594EA 262 EUR
BWRLBS2594
Anti-EDG-5 Rabbit Polyclonal Antibody [clone: L303]
Antikörper
Synthetic peptide, corresponding to amino acids 281-330 of Human EDG-5.

EDG-5 (L303) pAb detects endogenous levels of EDG-5 protein.

Type: Primary
Antigen: EDG-5
Clonality: Polyclonal
Clone: L303
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR