Anti-GPR173 Rabbit Polyclonal Antibody [clone: Y289]

Lieferant: Bioworld Technology
BS2610
BWRLBS2610EA 262 EUR
BWRLBS2610
Anti-GPR173 Rabbit Polyclonal Antibody [clone: Y289]
Antikörper
Synthetic peptide, corresponding to amino acids 256-310 of Human GPR173.

GPR173 (Y289) pAb detects endogenous levels of GPR173 protein.

Type: Primary
Antigen: GPR173
Clonality: Polyclonal
Clone: Y289
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR