Anti-Ku-86 Rabbit Polyclonal Antibody [clone: S462]
Lieferant: Bioworld Technology
BS2692
BWRLBS2692EA 262 EUR
BWRLBS2692
Anti-Ku-86 Rabbit Polyclonal Antibody [clone: S462]
Antikörper
Synthetic peptide, corresponding to amino acids 450-480 of Human Ku-86.
Ku-86 (S462) pAb detects endogenous levels of Ku-86 protein.
Type: Primary
Antigen: Ku-86
Clonality: Polyclonal
Clone: S462
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human