Anti-GPR153 Rabbit Polyclonal Antibody [clone: E234]

Lieferant: Bioworld Technology
BS2704
BWRLBS2704EA 262 EUR
BWRLBS2704
Anti-GPR153 Rabbit Polyclonal Antibody [clone: E234]
Antikörper
Synthetic peptide, corresponding to amino acids 201-256 of Human GPR153.

GPR153 (E234) pAb detects endogenous levels of GPR153 protein.

Type: Primary
Antigen: GPR153
Clonality: Polyclonal
Clone: E234
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR