Anti-mAChR M1 Rabbit Polyclonal Antibody [clone: R336]

Lieferant: Bioworld Technology
BS2708
BWRLBS2708EA 262 EUR
BWRLBS2708
Anti-mAChR M1 Rabbit Polyclonal Antibody [clone: R336]
Antikörper
Synthetic peptide, corresponding to amino acids 311-360 of Human mAChR M1.

mAChR M1 (R336) pAb detects endogenous levels of mAChR M1 protein.

Type: Primary
Antigen: mAChR M1
Clonality: Polyclonal
Clone: R336
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR