Anti-NMBR Rabbit Polyclonal Antibody [clone: L264]

Lieferant: Bioworld Technology
BS2737
BWRLBS2737EA 262 EUR
BWRLBS2737
Anti-NMBR Rabbit Polyclonal Antibody [clone: L264]
Antikörper
Synthetic peptide, corresponding to amino acids 235-280 of Human NMBR.

NMBR (L264) pAb detects endogenous levels of NMBR protein.

Type: Primary
Antigen: NMBR
Clonality: Polyclonal
Clone: L264
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR