Anti-MAGI-2 Rabbit Polyclonal Antibody [clone: E255]

Lieferant: Bioworld Technology
BS2754
BWRLBS2754EA 262 EUR
BWRLBS2754
Anti-MAGI-2 Rabbit Polyclonal Antibody [clone: E255]
Antikörper
Synthetic peptide, corresponding to amino acids 224-276 of Human MAGI-2.

MAGI-2 (E255) pAb detects endogenous levels of MAGI-2 protein.

Type: Primary
Antigen: MAGI-2
Clonality: Polyclonal
Clone: E255
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR