Anti-SR-5A Rabbit Polyclonal Antibody [clone: L35]

Lieferant: Bioworld Technology
BS2767
BWRLBS2767EA 262 EUR
BWRLBS2767
Anti-SR-5A Rabbit Polyclonal Antibody [clone: L35]
Antikörper
Synthetic peptide, corresponding to amino acids N-terminus of Human SR-5A.

SR-5A (L35) pAb detects endogenous levels of SR-5A protein.

Type: Primary
Antigen: SR-5A
Clonality: Polyclonal
Clone: L35
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR